Gene Transcripts

Gene Symbol
REEP2
Entrez Gene ID
51308
Gene Name
receptor accessory protein 2
Gene Aliases
C5orf19, SGC32445, SPG72, Yip2d
Chromosome Location
Chr. 5: 138439001 - 138446969 on Build GRCh38
UniGene ID
Hs.416090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271803.1 NP_001258732.1 8 2089
NM_016606.3 NP_057690.2 8 2083
NR_073448.1 8 2158
NR_073449.1 8 2164
GenBank mRNA AF070551.1 - 4 1527
AF251041.1 - 7 1863
AK056193.1 - 8 2045
AK223611.1 - 8 2030
BC006218.2 - 8 1997 3940709

View Full Product Details