Gene Transcripts

Gene Symbol
Gp1bb
Entrez Gene ID
116727
Gene Name
glycoprotein Ib (platelet), beta polypeptide
Gene Aliases
Chromosome Location
Chr. 11: 86520992 - 86522169 on Build Rnor_6.0
UniGene ID
Rn.96497
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053930.4 NP_446382.1 2 680
GenBank mRNA AB027146.1 - 1 1376
FQ227461.1 - 2 697

Gene Symbol
Sept5
Entrez Gene ID
116728
Gene Name
septin 5
Gene Aliases
5-Sep, CDCrel-1A, Pnutl1
Chromosome Location
Chr. 11: 86516390 - 86522169 on Build Rnor_6.0
UniGene ID
Rn.96497
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053931.3 NP_446383.3 11 3382
GenBank mRNA AB027143.1 - 11 3383

View Full Product Details