Gene Transcripts

Gene Symbol
AFTPH
Entrez Gene ID
54812
Gene Name
aftiphilin
Gene Aliases
Nbla10388
Chromosome Location
Chr. 2: 64524305 - 64593005 on Build GRCh38
UniGene ID
Hs.655167
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002243.2 NP_001002243.1 2 516
NM_017657.4 NP_060127.3 2 516
NM_203437.3 NP_982261.2 2 516
XM_005264380.1 XP_005264437.1 2 516
GenBank mRNA AK000087.1 - 2 491
AK222821.1 - 2 439
AL833962.2 - 2 294
AY367088.1 - 1 176
BC047529.1 - 2 490 5311682
BX504505.1 - 2 494
DA227019.1 - 2 505

View Full Product Details