Gene Transcripts

Gene Symbol
CNTLN
Entrez Gene ID
54875
Gene Name
centlein
Gene Aliases
C9orf101, C9orf39, bA340N12.1
Chromosome Location
Chr. 9: 17134991 - 17510019 on Build GRCh38
UniGene ID
Hs.435381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017738.3 NP_060208.2 8 1310
XM_005251492.1 XP_005251549.1 8 1310
XM_006716793.3 XP_006716856.1 8 1108
XM_011517941.2 XP_011516243.1 8 1310
XM_017014839.1 XP_016870328.1 8 1310
XM_017014840.1 XP_016870329.1 8 1310
XM_017014842.1 XP_016870331.1 8 1310
XM_017014843.1 XP_016870332.1 8 1310
GenBank mRNA AK000283.1 - 4 405
AK308837.1 - 8 1265
BX647069.1 - 8 1268

View Full Product Details