Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012013.1 | NP_001012013.1 | 9 | 1479 | |
XM_006247508.2 | XP_006247570.1 | 10 | 1473 | ||
XM_006247510.2 | XP_006247572.1 | 10 | 1468 | ||
XM_006247511.2 | XP_006247573.1 | 10 | 1464 | ||
XM_008768287.1 | XP_008766509.1 | 10 | 1477 | ||
GenBank mRNA | BC076387.1 | - | 9 | 1479 |