Gene Transcripts

Gene Symbol
TRNP1
Entrez Gene ID
388610
Gene Name
TMF1-regulated nuclear protein 1
Gene Aliases
C1orf225, TNRP
Chromosome Location
Chr. 1: 26993663 - 27000891 on Build GRCh38
UniGene ID
Hs.355747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013642.2 NP_001013664.1 2 1174
XM_005245867.3 XP_005245924.1 2 1233
GenBank mRNA BC069216.1 - 2 1171

View Full Product Details