Gene Transcripts

Gene Symbol
CATSPER4
Entrez Gene ID
378807
Gene Name
cation channel sperm associated 4
Gene Aliases
Chromosome Location
Chr. 1: 26189962 - 26202542 on Build GRCh38
UniGene ID
Hs.123532
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198137.1 NP_937770.1 9 1334
GenBank mRNA BC128138.1 - 9 1319
BC128139.1 - 9 1334 40112493
BC144663.1 - 8 1303

View Full Product Details