Gene Transcripts

Gene Symbol
Aym1
Entrez Gene ID
503692
Gene Name
activator of yeast meiotic promoters 1
Gene Aliases
Chromosome Location
Chr. 5: 113357294 - 113357821 on Build GRCm38
UniGene ID
Mm.348794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012726.2 NP_001012744.2 1 264
GenBank mRNA AF069744.1 - 1 270
BC145860.1 - 1 124 40131163
BC145862.1 - 1 124 40131165

View Full Product Details