Gene Transcripts
- Gene Symbol
- ATAD3B
- Entrez Gene ID
- 83858
- Gene Name
- ATPase family, AAA domain containing 3B
- Gene Aliases
- AAA-TOB3, TOB3
- Chromosome Location
- Chr. 1: 1471732 - 1509466 on Build GRCh38
- UniGene ID
- Hs.729021
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317238.1 |
NP_001304167.1
|
10, 11 |
1516 |
|
NM_031921.5 |
NP_114127.3
|
12, 13 |
1399 |
|
XM_005244806.3 |
XP_005244863.1
|
12, 13 |
1422 |
|
XM_011542241.2 |
XP_011540543.1
|
12, 13 |
1422 |
|
XM_011542243.2 |
XP_011540545.1
|
11, 12 |
1135 |
|
XM_011542244.1 |
XP_011540546.1
|
12, 13 |
1566 |
|
GenBank mRNA |
AB033099.1 |
- |
10, 11 |
1605 |
|
AF343078.1 |
- |
12, 13 |
1332 |
|
AK024028.1 |
- |
10, 11 |
984 |
|
AK127857.1 |
- |
5, 6 |
2050 |
|
AK290586.1 |
- |
12, 13 |
1391 |
|
BC002542.2 |
- |
12, 13 |
1325 |
3138578 |
BC018701.1 |
- |
12, 13 |
1325 |
3139886 |
View Full Product Details