Gene Transcripts

Gene Symbol
Brox
Entrez Gene ID
71678
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
0610010K06Rik
Chromosome Location
Chr. 1: 183276342 - 183297274 on Build GRCm38
UniGene ID
Mm.392270
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027861.2 NP_082137.1 4, 5 426
GenBank mRNA AK002489.1 - 4, 5 396
AK050525.1 - 4, 5 397
AK162247.1 - 4, 5 396
BB857916.1 - 4, 5 428
BC030340.1 - 4, 5 432 3498026
BC120692.1 - 3, 4 274 40129615
BC125527.1 - 3, 4 274 40130042

View Full Product Details