Gene Transcripts

Gene Symbol
Mnda
Entrez Gene ID
304988
Gene Name
myeloid cell nuclear differentiation antigen
Gene Aliases
Ifi204, rHin-3
Chromosome Location
Chr. 13: 92073664 - 92091335 on Build Rnor_6.0
UniGene ID
Rn.15194
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012029.1 NP_001012029.1 6 844
GenBank mRNA BC085891.1 - 6 844

View Full Product Details