Gene Transcripts

Gene Symbol
Cage1
Entrez Gene ID
306872
Gene Name
cancer antigen 1
Gene Aliases
Ctag3
Chromosome Location
Chr. 17: 27452506 - 27490853 on Build Rnor_6.0
UniGene ID
Rn.128696
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012052.1 NP_001012052.1 9 2490
XM_006253808.2 XP_006253870.1 9 2477
XM_006253809.2 XP_006253871.1 8 2168
XM_006253810.2 XP_006253872.1 8 2360
XM_006253811.2 XP_006253873.1 8 2444
GenBank mRNA BC081934.1 - 9 2490
FQ211932.1 - 8 2448

View Full Product Details