Gene Transcripts

Gene Symbol
Bfar
Entrez Gene ID
304709
Gene Name
bifunctional apoptosis regulator
Gene Aliases
Chromosome Location
Chr. 10: 1464783 - 1497653 on Build Rnor_6.0
UniGene ID
Rn.159939
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013125.1 NP_001013143.1 8 2288
GenBank mRNA BC087103.1 - 8 2288

View Full Product Details