Gene Transcripts

Gene Symbol
Mybphl
Entrez Gene ID
310782
Gene Name
myosin binding protein H-like
Gene Aliases
RGD1310511
Chromosome Location
Chr. 2: 211158917 - 211173036 on Build Rnor_6.0
UniGene ID
Rn.139442
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014042.1 NP_001014064.1 9 1268
XM_006233150.2 XP_006233212.1 9 2662
GenBank mRNA BC087113.1 - 9 1268

View Full Product Details