Gene Transcripts

Gene Symbol
Nrros
Entrez Gene ID
303875
Gene Name
negative regulator of reactive oxygen species
Gene Aliases
Garpl1, Lrrc33, RGD1310771
Chromosome Location
Chr. 11: 71872729 - 71889897 on Build Rnor_6.0
UniGene ID
Rn.138732
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024995.1 NP_001020166.1 3 2195
XM_006248448.2 XP_006248510.1 3 2236
GenBank mRNA BC091190.1 - 3 2195

View Full Product Details