Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002231.2 | NP_001002231.1 | 5 | 1939 | |
NM_001256080.1 | NP_001243009.1 | 4 | 1742 | ||
NM_005551.4 | NP_005542.1 | 5 | 1902 | ||
NR_045762.1 | 5 | 1926 | |||
NR_045763.1 | 5 | 1988 | |||
GenBank mRNA | AF188745.1 | - | 5 | 1901 | |
AF188746.1 | - | 5 | 1877 | ||
AF188747.1 | - | 5 | 1914 |