Gene Transcripts

Gene Symbol
STXBP4
Entrez Gene ID
252983
Gene Name
syntaxin binding protein 4
Gene Aliases
Synip
Chromosome Location
Chr. 17: 54968728 - 55164285 on Build GRCh38
UniGene ID
Hs.35199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178509.5 NP_848604.3 3, 4 246
XM_005257187.4 XP_005257244.1 3, 4 282
XM_006721797.3 XP_006721860.1 3, 4 280
XM_006721798.3 XP_006721861.1 3, 4 277
XM_017024410.1 XP_016879899.1 3, 4 283
XM_017024411.1 XP_016879900.1 3, 4 281
XM_017024412.1 XP_016879901.1 3, 4 280
XM_017024414.1 XP_016879903.1 3, 4 282
XM_017024415.1 XP_016879904.1 3, 4 282
GenBank mRNA AK122865.1 - 3, 4 246
BC125116.1 - 3, 4 235 40118892

View Full Product Details