Gene Transcripts
- Gene Symbol
- IQCC
- Entrez Gene ID
- 55721
- Gene Name
- IQ motif containing C
- Gene Aliases
- Chromosome Location
- Chr. 1: 32205635 - 32208687 on Build GRCh38
- UniGene ID
- Hs.274356
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001160042.1 |
NP_001153514.1
|
2, 3 |
484 |
|
NM_018134.2 |
NP_060604.2
|
2, 3 |
244 |
|
GenBank mRNA |
AK001409.1 |
- |
2, 3 |
208 |
|
AK223272.1 |
- |
2, 3 |
232 |
|
AK299235.1 |
- |
2 |
419 |
|
AK307853.1 |
- |
2, 3 |
448 |
|
BC098250.1 |
- |
2, 3 |
207 |
|
BC098260.1 |
- |
2, 3 |
207 |
40024457 |
BC098317.1 |
- |
2, 3 |
207 |
40024460 |
BC098366.1 |
- |
2, 3 |
207 |
40024462 |
DA746781.1 |
- |
2, 3 |
244 |
|
View Full Product Details