Gene Transcripts

Gene Symbol
SLC38A8
Entrez Gene ID
146167
Gene Name
solute carrier family 38 member 8
Gene Aliases
FVH2
Chromosome Location
Chr. 16: 84009667 - 84043372 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080442.2 NP_001073911.1 7 857
XM_017022946.1 XP_016878435.1 9 1322

View Full Product Details