Gene Transcripts

Gene Symbol
SMIM24
Entrez Gene ID
284422
Gene Name
small integral membrane protein 24
Gene Aliases
C19orf77, HSPC323
Chromosome Location
Chr. 19: 3474407 - 3480542 on Build GRCh38
UniGene ID
Hs.130714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136503.1 NP_001129975.1 4 897
GenBank mRNA AF161441.1 - 3 753

View Full Product Details