Gene Transcripts

Gene Symbol
PRR34
Entrez Gene ID
55267
Gene Name
proline rich 34
Gene Aliases
C22orf26
Chromosome Location
Chr. 22: 46050459 - 46054144 on Build GRCh38
UniGene ID
Hs.567529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018280.2 NP_060750.1 1, 2 402
GenBank mRNA AK001807.1 - 1, 2 402
AK096201.1 - 1, 2 54
AU134173.1 - 1, 2 402

View Full Product Details