Gene Transcripts

Gene Symbol
MRGBP
Entrez Gene ID
55257
Gene Name
MRG/MORF4L binding protein
Gene Aliases
C20orf20, Eaf7, MRG15BP, URCC4
Chromosome Location
Chr. 20: 62796405 - 62801738 on Build GRCh38
UniGene ID
Hs.443260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018270.5 NP_060740.1 3 435
NR_136405.1 3 435
GenBank mRNA AB085682.1 - 3 335
AK001776.1 - 3 387
BC009889.2 - 3 354 4026378
BC014235.2 - 3 359 4555060
BC018841.2 - 3 354 3050866
BM920599.1 - 341
BU902487.1 - 3 417

View Full Product Details