Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017927.3 | NP_001017927.2 | 7 | 1030 | |
NM_001322329.1 | NP_001309258.1 | 7 | 951 | ||
NM_001322330.1 | NP_001309259.1 | 6 | 827 | ||
NM_001322331.1 | NP_001309260.1 | 6 | 480 | ||
NM_001322332.1 | NP_001309261.1 | 5 | 429 | ||
XM_017003354.1 | XP_016858843.1 | 7 | 628 | ||
GenBank mRNA | BC047365.1 | - | 7 | 951 | |
BC062434.1 | - | 5 | 462 | ||
BC105805.1 | - | 6 | 484 | 6496685 |