Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
56942
Gene Name
C-X9-C motif containing 2
Gene Aliases
2310061C15Rik, C16orf61, DC13
Chromosome Location
Chr. 16: 80975800 - 81007046 on Build GRCh38
UniGene ID
Hs.55028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020188.3 NP_064573.1 2, 3 273
XM_017023463.1 XP_016878952.1 2, 3 401
XM_017023467.1 XP_016878956.1 4, 5 356
XM_017023468.1 XP_016878957.1 3, 4 266
XM_017023469.1 XP_016878958.1 3, 4 340
XM_017023470.1 XP_016878959.1 3, 4 264
GenBank mRNA AF201935.1 - 2, 3 249
AK123193.1 - 1, 2 1628
BC032631.1 - 2, 3 277 5399193

View Full Product Details