Gene Transcripts

Gene Symbol
Ly49s7
Entrez Gene ID
494203
Gene Name
Ly49 stimulatory receptor 7
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.129029
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009494.1 NP_001009494.1 2 161
GenBank mRNA AY649833.1 - 2 161
AY649840.1 - 2 153

Gene Symbol
LOC502908
Entrez Gene ID
502908
Gene Name
Ly49s8
Gene Aliases
Chromosome Location
Chr. 4: 165093769 - 165118489 on Build Rnor_6.0
UniGene ID
Rn.145852
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763439.1 XP_008761661.1 1 444
GenBank mRNA AY669088.1 - 1 17

View Full Product Details