Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098845.2 | NP_001092315.2 | 3 | 396 | |
NM_001278923.1 | NP_001265852.1 | 3 | 396 | ||
NM_001278924.1 | NP_001265853.1 | 3 | 510 | ||
GenBank mRNA | AK223149.1 | - | 3 | 210 | |
AK300191.1 | - | 3 | 230 | ||
AK307182.1 | - | 3 | 210 | ||
AK313626.1 | - | 3 | 259 | ||
BC008813.2 | - | 3 | 424 | 3958020 | |
DA698972.1 | - | 3 | 378 | ||
DC381265.1 | - | 3 | 397 | ||
X16662.1 | - | 3 | 240 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040084.2 | NP_001035173.1 | 3 | 359 | |
NM_001271702.1 | NP_001258631.1 | 3 | 473 | ||
XM_006717951.3 | XP_006718014.1 | 3 | 313 | ||
XM_011540096.1 | XP_011538398.1 | 4 | 512 | ||
XM_011540098.1 | XP_011538400.1 | 4 | 512 | ||
XM_011540099.1 | XP_011538401.1 | 4 | 512 | ||
XM_011540101.2 | XP_011538403.1 | 4 | 512 | ||
GenBank mRNA | AK296976.1 | - | 3 | 263 | |
AK298761.1 | - | 3 | 339 | ||
AK300139.1 | - | 3 | 324 | ||
AK300158.1 | - | 3 | 217 | ||
BC004376.1 | - | 3 | 256 | 3639064 | |
BC073755.1 | - | 3 | 290 | 6538697 | |
CB270161.1 | - | 3 | 345 | ||
DC339657.1 | - | 3 | 359 | ||
M81844.1 | - | 3 | 161 | ||
Z36830.1 | - | 3 | 270 |