Gene Transcripts

Gene Symbol
Aimp1
Entrez Gene ID
114632
Gene Name
aminoacyl tRNA synthetase complex-interacting multifunctional protein 1
Gene Aliases
Emap2, Scye1
Chromosome Location
Chr. 2: 237727782 - 237751327 on Build Rnor_6.0
UniGene ID
Rn.3192
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053757.1 NP_446209.1 6 735
GenBank mRNA AF320763.1 - 2 159
BC098675.1 - 6 735 7322792
FQ226969.1 - 4 511
FQ229959.1 - 4 511

View Full Product Details