Gene Transcripts

Gene Symbol
Ubfd1
Entrez Gene ID
293454
Gene Name
ubiquitin family domain containing 1
Gene Aliases
RGD1306614, Ubph
Chromosome Location
Chr. 1: 192025437 - 192040506 on Build Rnor_6.0
UniGene ID
Rn.198517
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034911.1 NP_001030083.1 6 805
XM_006230189.2 XP_006230251.1 7 1149
GenBank mRNA BC101866.1 - 6 805

View Full Product Details