Gene Transcripts

Gene Symbol
Adam4
Entrez Gene ID
57022
Gene Name
a disintegrin and metalloprotease domain 4
Gene Aliases
tMDCV
Chromosome Location
Chr. 6: 105158024 - 105160470 on Build Rnor_6.0
UniGene ID
Rn.209651
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020305.1 NP_064701.1 1 244
GenBank mRNA BC107437.1 - 1 244

View Full Product Details