Gene Transcripts

Gene Symbol
RSRP1
Entrez Gene ID
57035
Gene Name
arginine and serine rich protein 1
Gene Aliases
C1orf63, NPD014
Chromosome Location
Chr. 1: 25242237 - 25338254 on Build GRCh38
UniGene ID
Hs.259412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321772.1 NP_001308701.1 4 1973
NM_020317.4 NP_064713.3 4 969
NR_135143.1 3 2485
NR_135144.1 4 1560
NR_135777.1 3 2460
NR_135778.1 4 1834
NR_135780.1 4 1938
NR_135781.1 5 1605
NR_135782.1 5 1287
NR_135783.1 5 1014
NR_135784.1 3 2485
NR_135785.1 5 1013
NR_135786.1 3 2485
NR_135787.1 5 2609
NR_135788.1 3 2551
NR_135789.1 3 3489
XM_011541797.1 XP_011540099.1 4 934
GenBank mRNA AF267856.1 - 3 2413
AK226012.1 - 4 1497
AK292532.1 - 3 2421
BC001892.1 - 4 893 3534629
BC041843.1 - 5 2520 5269181
BC065040.1 - 4 882 6147639

View Full Product Details