Gene Transcripts
- Gene Symbol
- Vmn1r64
- Entrez Gene ID
- 404285
- Gene Name
- vomeronasal 1 receptor 64
- Gene Aliases
- V1rd11
- Chromosome Location
- Chr. 7: 5883580 - 5884542 on Build GRCm38
- UniGene ID
- Mm.377929
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207544.1 |
NP_997427.1
|
1 |
650 |
|
View Full Product Details