Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033537.2 | NP_001028709.2 | 7 | 867 | |
GenBank mRNA | AK052486.1 | - | 7 | 784 | |
AK078974.1 | - | 7 | 793 | ||
AK170817.1 | - | 7 | 782 | ||
BC030368.1 | - | 5 | 487 | ||
BC035945.1 | - | 7 | 790 |