Gene Transcripts

Gene Symbol
RSRP1
Entrez Gene ID
57035
Gene Name
arginine and serine rich protein 1
Gene Aliases
C1orf63, NPD014
Chromosome Location
Chr. 1: 25242237 - 25338254 on Build GRCh38
UniGene ID
Hs.259412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321772.1 NP_001308701.1 5 2377
NM_020317.4 NP_064713.3 5 1373
NR_135143.1 4 2889
NR_135144.1 5 1964
NR_135777.1 4 2864
NR_135778.1 5 2238
NR_135780.1 5 2342
NR_135781.1 6 2009
NR_135782.1 6 1691
NR_135783.1 6 1418
NR_135784.1 5 2777
NR_135785.1 6 1417
NR_135786.1 5 2764
NR_135787.1 6 3013
NR_135788.1 4 2955
NR_135789.1 4 3893
XM_011541797.1 XP_011540099.1 5 1338
GenBank mRNA AF267856.1 - 4 2817
AK226012.1 - 5 1901
AK292532.1 - 4 2825
BC001892.1 - 5 1297 3534629
BC041843.1 - 6 2924 5269181
BC065040.1 - 5 1286 6147639
BM875537.1 - 2 408

View Full Product Details