Gene Transcripts
- Gene Symbol
- ANKRD26
- Entrez Gene ID
- 22852
- Gene Name
- ankyrin repeat domain 26
- Gene Aliases
- THC2, bA145E8.1
- Chromosome Location
- Chr. 10: 26938195 - 27100498 on Build GRCh38
- UniGene ID
- Hs.361041
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256053.1 |
NP_001242982.1
|
15, 16 |
1741 |
|
NM_014915.2 |
NP_055730.2
|
15, 16 |
1741 |
|
XM_006717423.2 |
XP_006717486.1
|
15, 16 |
1741 |
|
XM_006717425.3 |
XP_006717488.1
|
15, 16 |
1741 |
|
XM_011519416.2 |
XP_011517718.1
|
15, 16 |
1741 |
|
XM_017015928.1 |
XP_016871417.1
|
15, 16 |
1741 |
|
XM_017015929.1 |
XP_016871418.1
|
16, 17 |
1729 |
|
XM_017015930.1 |
XP_016871419.1
|
15, 16 |
1741 |
|
XM_017015931.1 |
XP_016871420.1
|
15, 16 |
1741 |
|
XM_017015932.1 |
XP_016871421.1
|
15, 16 |
1741 |
|
XM_017015933.1 |
XP_016871422.1
|
15, 16 |
1741 |
|
XM_017015934.1 |
XP_016871423.1
|
15, 16 |
1741 |
|
GenBank mRNA |
AB028997.1 |
- |
15, 16 |
1719 |
|
AK001137.1 |
- |
15, 16 |
1735 |
|
BC130453.1 |
- |
15, 16 |
1682 |
40146484 |
View Full Product Details