Gene Transcripts

Gene Symbol
SPDYE3
Entrez Gene ID
441272
Gene Name
speedy/RINGO cell cycle regulator family member E3
Gene Aliases
Chromosome Location
Chr. 7: 100306799 - 100322196 on Build GRCh38
UniGene ID
Hs.703626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004351.4 NP_001004351.3 11 3086
GenBank mRNA BC056606.1 - 11 3087

View Full Product Details