Gene Transcripts
- Gene Symbol
- Smagp
- Entrez Gene ID
- 207818
- Gene Name
- small cell adhesion glycoprotein
- Gene Aliases
- Chromosome Location
- Chr. 15: 100621339 - 100636940 on Build GRCm38
- UniGene ID
- Mm.26282
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033872.2 |
NP_001029044.1
|
2, 3 |
257 |
|
NM_174992.4 |
NP_778157.2
|
2, 3 |
447 |
|
GenBank mRNA |
BC004728.1 |
- |
2, 3 |
248 |
|
BC132460.1 |
- |
1, 2 |
54 |
40130737 |
BC132462.1 |
- |
1, 2 |
54 |
40130739 |
BE132795.1 |
- |
2, 3 |
181 |
|
BF018035.1 |
- |
2, 3 |
325 |
|
BG089628.1 |
- |
2, 3 |
362 |
|
BY707988.1 |
- |
2, 3 |
182 |
|
CJ042753.1 |
- |
2, 3 |
210 |
|
CJ053067.1 |
- |
2, 3 |
260 |
|
CJ065989.1 |
- |
2, 3 |
359 |
|
CK129185.1 |
- |
2, 3 |
151 |
|
View Full Product Details