Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029751.4 | NP_084027.1 | 4 | 459 | |
GenBank mRNA | AK010983.1 | - | 4 | 430 | |
AK146543.1 | - | 4 | 429 | ||
AK150820.1 | - | 4 | 428 | ||
AK152611.1 | - | 4 | 429 | ||
AK161942.1 | - | 4 | 429 | ||
AK162047.1 | - | 4 | 430 | ||
AK168258.1 | - | 4 | 429 | ||
BC024609.1 | - | 4 | 403 | 4221597 | |
BC037146.1 | - | 4 | 471 | 5011713 | |
CJ042325.1 | - | 4 | 461 |