Gene Transcripts

Gene Symbol
WDR35
Entrez Gene ID
57539
Gene Name
WD repeat domain 35
Gene Aliases
CED2, IFT121, IFTA1, SRTD7
Chromosome Location
Chr. 2: 19910260 - 19990123 on Build GRCh38
UniGene ID
Hs.205427
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006657.1 NP_001006658.1 10 1238
NM_020779.3 NP_065830.2 10 1238
XM_011533007.2 XP_011531309.1 1 24
GenBank mRNA AB037757.2 - 10 1246
AK091205.1 - 5 667
AK122917.1 - 10 1163
AK315094.1 - 10 1163
BC036659.2 - 10 1237 5266940

View Full Product Details