Gene Transcripts

Gene Symbol
ZFAND3
Entrez Gene ID
60685
Gene Name
zinc finger AN1-type containing 3
Gene Aliases
TEX27
Chromosome Location
Chr. 6: 37819496 - 38154624 on Build GRCh38
UniGene ID
Hs.36959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021943.2 NP_068762.1 5 855
XM_005249269.1 XP_005249326.1 4 824
XM_017011171.1 XP_016866660.1 4 828
GenBank mRNA AK023284.1 - 5 529
BC031481.2 - 5 471 4448968

View Full Product Details