Gene Transcripts
- Gene Symbol
- GLIPR2
- Entrez Gene ID
- 152007
- Gene Name
- GLI pathogenesis related 2
- Gene Aliases
- C9orf19, GAPR-1, GAPR1
- Chromosome Location
- Chr. 9: 36136536 - 36163913 on Build GRCh38
- UniGene ID
- Hs.493819
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287011.1 |
NP_001273940.1
|
3, 4 |
477 |
|
NM_001287013.1 |
NP_001273942.1
|
3, 4 |
314 |
|
NM_022343.3 |
NP_071738.1
|
3, 4 |
477 |
|
NR_104637.1 |
|
5, 6 |
858 |
|
NR_104638.1 |
|
3, 4 |
269 |
|
NR_104639.1 |
|
3, 4 |
477 |
|
NR_104640.1 |
|
3, 4 |
499 |
|
GenBank mRNA |
AJ011129.1 |
- |
2, 3 |
234 |
|
AK074488.1 |
- |
3, 4 |
299 |
|
AK075091.1 |
- |
3, 4 |
278 |
|
AK091288.1 |
- |
4, 5 |
1091 |
|
AK298741.1 |
- |
3, 4 |
278 |
|
AK307243.1 |
- |
3, 4 |
278 |
|
AY039756.1 |
- |
3, 4 |
268 |
|
BC017918.1 |
- |
3, 4 |
280 |
4296721 |
CR749781.1 |
- |
5, 6 |
659 |
|
DA268281.1 |
- |
3, 4 |
314 |
|
DA271525.1 |
- |
3, 4 |
254 |
|
DA824243.1 |
- |
3, 4 |
268 |
|
DA986727.1 |
- |
3, 4 |
278 |
|
View Full Product Details