Gene Transcripts

Gene Symbol
RIC3
Entrez Gene ID
79608
Gene Name
RIC3 acetylcholine receptor chaperone
Gene Aliases
AYST720, PRO1385
Chromosome Location
Chr. 11: 8092965 - 8169088 on Build GRCh38
UniGene ID
Hs.231850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206671.2 NP_001193600.1 1, 2 186
NM_001206672.2 NP_001193601.1 1, 2 186
NM_024557.4 NP_078833.3 1, 2 186
NR_045405.1 2, 3 235
XM_006718317.3 XP_006718380.1 1, 2 231
XM_006718318.3 XP_006718381.1 1, 2 231
XM_006718321.3 XP_006718384.1 1, 2 231
XM_017018287.1 XP_016873776.1 1, 2 231
XM_017018288.1 XP_016873777.1 2, 3 228
XM_017018289.1 XP_016873778.1 2, 3 262
XM_017018290.1 XP_016873779.1 1, 2 231
GenBank mRNA AK293933.1 - 1, 2 149
AK296505.1 - 2, 3 200
AK315379.1 - 1, 2 145
AM422214.1 - 1, 2 132
AY326435.1 - 1, 2 132
AY326436.1 - 1, 2 167
AY358475.1 - 1, 2 157
BC022455.1 - 1, 2 168 4792930
DC420812.1 - 1, 2 186
KU178773.1 - 1, 2 132
KU178774.1 - 1, 2 132

View Full Product Details