Gene Transcripts
- Gene Symbol
- C11orf63
- Entrez Gene ID
- 79864
- Gene Name
- chromosome 11 open reading frame 63
- Gene Aliases
- Chromosome Location
- Chr. 11: 122881640 - 122963862 on Build GRCh38
- UniGene ID
- Hs.164705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024806.3 |
NP_079082.2
|
2, 3 |
886 |
|
NM_199124.2 |
NP_954575.1
|
2, 3 |
886 |
|
XM_005271680.4 |
XP_005271737.1
|
2, 3 |
747 |
|
XM_006718915.2 |
XP_006718978.1
|
2, 3 |
621 |
|
XM_011543009.2 |
XP_011541311.1
|
2, 3 |
955 |
|
XM_017018338.1 |
XP_016873827.1
|
2, 3 |
621 |
|
XM_017018339.1 |
XP_016873828.1
|
2, 3 |
654 |
|
XM_017018340.1 |
XP_016873829.1
|
2, 3 |
465 |
|
XM_017018341.1 |
XP_016873830.1
|
2, 3 |
1236 |
|
XM_017018342.1 |
XP_016873831.1
|
2, 3 |
621 |
|
XM_017018343.1 |
XP_016873832.1
|
2, 3 |
621 |
|
GenBank mRNA |
AK027207.1 |
- |
1, 2 |
240 |
|
AK097534.1 |
- |
2, 3 |
731 |
|
AK291625.1 |
- |
2, 3 |
489 |
|
BC009820.2 |
- |
1, 2 |
292 |
|
BC068507.1 |
- |
2, 3 |
649 |
5298472 |
View Full Product Details