Gene Transcripts
- Gene Symbol
- ACSF2
- Entrez Gene ID
- 80221
- Gene Name
- acyl-CoA synthetase family member 2
- Gene Aliases
- ACSMW, AVYV493
- Chromosome Location
- Chr. 17: 50426158 - 50474845 on Build GRCh38
- UniGene ID
- Hs.288959
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288968.1 |
NP_001275897.1
|
3, 4 |
505 |
|
NM_001288969.1 |
NP_001275898.1
|
2, 3 |
430 |
|
NM_001288972.1 |
NP_001275901.1
|
2, 3 |
430 |
|
NM_025149.5 |
NP_079425.3
|
2, 3 |
430 |
|
XM_006722110.3 |
XP_006722173.1
|
2, 3 |
370 |
|
XM_011525294.1 |
XP_011523596.1
|
2, 3 |
370 |
|
GenBank mRNA |
AK024573.1 |
- |
2, 3 |
337 |
|
AK225044.1 |
- |
2, 3 |
337 |
|
AK225561.1 |
- |
2, 3 |
336 |
|
AK294205.1 |
- |
3, 4 |
447 |
|
AK295258.1 |
- |
2, 3 |
338 |
|
AY358660.1 |
- |
2, 3 |
366 |
|
BC012053.1 |
- |
2, 3 |
343 |
4577089 |
BC014123.2 |
- |
2, 3 |
350 |
4560029 |
DC296100.1 |
- |
2, 3 |
370 |
|
View Full Product Details