Gene Transcripts
- Gene Symbol
- ACSF2
- Entrez Gene ID
- 80221
- Gene Name
- acyl-CoA synthetase family member 2
- Gene Aliases
- ACSMW, AVYV493
- Chromosome Location
- Chr. 17: 50426158 - 50474845 on Build GRCh38
- UniGene ID
- Hs.288959
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288968.1 |
NP_001275897.1
|
16, 17 |
1979 |
|
NM_001288969.1 |
NP_001275898.1
|
15, 16 |
1865 |
|
NM_001288970.1 |
NP_001275899.1
|
14, 15 |
1775 |
|
NM_001288971.1 |
NP_001275900.1
|
13, 14 |
1579 |
|
NM_001288972.1 |
NP_001275901.1
|
15, 16 |
1821 |
|
NM_025149.5 |
NP_079425.3
|
15, 16 |
1904 |
|
NR_110232.1 |
|
5, 6 |
537 |
|
XM_011525294.1 |
XP_011523596.1
|
15, 16 |
1826 |
|
GenBank mRNA |
AK024573.1 |
- |
15, 16 |
1811 |
|
AK225044.1 |
- |
15, 16 |
1811 |
|
AK225561.1 |
- |
15, 16 |
1810 |
|
AK294205.1 |
- |
16, 17 |
1921 |
|
AK295258.1 |
- |
15, 16 |
1773 |
|
AK300145.1 |
- |
14, 15 |
1683 |
|
AK300625.1 |
- |
13, 14 |
1579 |
|
AK308584.1 |
- |
5, 6 |
523 |
|
AY358660.1 |
- |
15, 16 |
1840 |
|
BC012053.1 |
- |
15, 16 |
1817 |
4577089 |
BC014123.2 |
- |
15, 16 |
1824 |
4560029 |
View Full Product Details