Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161009.2 | NP_001154481.1 | 2 | 622 | |
NM_001161010.2 | NP_001154482.1 | 3 | 727 | ||
NM_001161011.2 | NP_001154483.1 | 2 | 622 | ||
NM_001161012.2 | NP_001154484.1 | 2 | 622 | ||
NM_001161013.2 | NP_001154485.1 | 2 | 622 | ||
NM_031905.4 | NP_114111.2 | 3 | 727 | ||
XM_005250628.2 | XP_005250685.1 | 3 | 881 | ||
XM_011516601.2 | XP_011514903.1 | 2 | 800 | ||
XM_011516602.2 | XP_011514904.1 | 2 | 161 | ||
XM_017012684.1 | XP_016868173.1 | 4 | 908 | ||
XM_017012685.1 | XP_016868174.1 | 4 | 908 | ||
XM_017012686.1 | XP_016868175.1 | 4 | 908 | ||
XM_017012687.1 | XP_016868176.1 | 2 | 140 | ||
XM_017012688.1 | XP_016868177.1 | 4 | 908 | ||
XM_017012689.1 | XP_016868178.1 | 4 | 908 | ||
GenBank mRNA | AF275808.1 | - | 2 | 130 | |
AK075500.1 | - | 3 | 468 | ||
AK225082.1 | - | 2 | 355 | ||
AK291818.1 | - | 2 | 350 | ||
AK301565.1 | - | 2 | 355 | ||
AY150851.1 | - | 2 | 622 | ||
AY150852.1 | - | 3 | 727 | ||
AY150853.1 | - | 2 | 622 | ||
AY150854.1 | - | 3 | 727 | ||
BC003586.1 | - | 2 | 342 | 3357276 |