Gene Transcripts

Gene Symbol
EVA1C
Entrez Gene ID
59271
Gene Name
eva-1 homolog C (C. elegans)
Gene Aliases
B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21
Chromosome Location
Chr. 21: 32411692 - 32515400 on Build GRCh38
UniGene ID
Hs.208358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286556.1 NP_001273485.1 7 1312
NM_001320744.1 NP_001307673.1 5 1056
NM_001320745.1 NP_001307674.1 7 1246
NM_058187.4 NP_478067.2 7 1321
NR_104472.1 6 1115
NR_135467.1 7 1420
XM_006724038.3 XP_006724101.1 7 1439
XM_011529669.2 XP_011527971.1 7 1573
XM_017028418.1 XP_016883907.1 7 1439
XM_017028419.1 XP_016883908.1 6 1361
XM_017028420.1 XP_016883909.1 8 1362
XM_017028421.1 XP_016883910.1 7 1237
XM_017028422.1 XP_016883911.1 5 1112
XM_017028423.1 XP_016883912.1 4 1040
GenBank mRNA AF358258.1 - 7 904
AK125013.1 - 6 1871
AW957938.1 - 2 133
AY040087.1 - 6 707
AY358787.1 - 7 1314
BC038710.1 - 7 1047 4813873

View Full Product Details