Gene Transcripts

Gene Symbol
GNG5
Entrez Gene ID
2787
Gene Name
G protein subunit gamma 5
Gene Aliases
Chromosome Location
Chr. 1: 84498323 - 84506579 on Build GRCh38
UniGene ID
Hs.645427
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005274.2 NP_005265.1 3, 4 588
GenBank mRNA AF038955.1 - 2, 3 271
BC003563.1 - 2, 3 317 3502879
BM809260.1 - 2, 3 327

View Full Product Details