Gene Transcripts

Gene Symbol
C4orf22
Entrez Gene ID
255119
Gene Name
chromosome 4 open reading frame 22
Gene Aliases
Chromosome Location
Chr. 4: 80321265 - 80963756 on Build GRCh38
UniGene ID
Hs.527104
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206997.1 NP_001193926.1 2 251
NM_152770.2 NP_689983.2 2 251
XM_011531816.2 XP_011530118.2 3 417
XM_017007972.1 XP_016863461.1 3 403
XM_017007973.1 XP_016863462.1 3 406
XM_017007974.1 XP_016863463.1 3 411
XM_017007976.1 XP_016863465.1 3 426
XM_017007977.1 XP_016863466.1 3 417
GenBank mRNA AK128642.1 - 2 203
BC034296.1 - 2 251 5165558

View Full Product Details