Gene Transcripts

Gene Symbol
SPATA33
Entrez Gene ID
124045
Gene Name
spermatogenesis associated 33
Gene Aliases
C16orf55
Chromosome Location
Chr. 16: 89657744 - 89670458 on Build GRCh38
UniGene ID
Hs.655171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271907.1 NP_001258836.1 3 358
NM_001271908.1 NP_001258837.1 3 276
NM_001271909.1 NP_001258838.1 2 414
NM_001271910.1 NP_001258839.1 3 356
NM_153025.2 NP_694570.1 3 305
NR_073515.1 2 445
GenBank mRNA AK056168.1 - 3 247
AK125673.1 - 2 186
AK128803.1 - 2 420
AK303024.1 - 3 324
BC148299.2 - 3 247 40112476
BP345094.1 - 2 414
DA300380.1 - 3 268
DB025192.1 - 3 305
DB452323.1 - 3 358
DC357681.1 - 3 314
DC374690.1 - 3 326

View Full Product Details