Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_026542.1 | 2 | 591 | ||
NR_026543.1 | 2 | 591 | |||
GenBank mRNA | AF426266.1 | - | 2 | 210 | |
AF426267.1 | - | 2 | 210 | ||
BC080530.1 | - | 2 | 590 | ||
BC119737.2 | - | 2 | 184 | ||
BC119738.2 | - | 2 | 184 | ||
BG751912.1 | - | 2 | 590 | ||
DA218783.1 | - | 2 | 370 |